Learn More
Electron Microscopy Sciences Aurion ImmunoGold EM Gar-6/Gam-10/BSA-C Kit
SDP
Supplier:  Electron Microscopy Sciences 25540
Encompass_Preferred
Immunogold reagent kit. Two non-overlapping gold sizes coupled to either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies. Includes: 1ml Goat-anti-Rabbit 6 nm, 1ml Goat-anti-Mouse IgG/IgM 10 nm, 30ml BSA-c
Catalog No. 50-282-88