Learn More
Electron Microscopy Sciences Aurion ImmunoGold EM Gam-6/Gar-10.Bsa-C Kit
SDP
Supplier:  Electron Microscopy Sciences 25544
Encompass_Preferred
Immunogold reagent kit. Two non-overlapping gold particle sizes. Coupled to either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies, includes: 1ml Goat-anti-Mouse IgG/IgM 6 nm, 1ml Goat-anti-Rabbit 10 nm, 30ml BSA-c
Catalog No. 50-282-94