Learn More
Alkali Scientific ASI™ Immunoplate (ELISA) Strip Single Well Plate, PS, Flat Bottom, 8 Well, 100/Cs, Maxibinding
SDP
Supplier:  Alkali Scientific SSWMAXI
Encompass_Preferred
▫ Multibinding : Suitable specifically for adsorbing hydrophilic molecules. (Antigen ELISA)▫ Maxibinding : Modified polystyrene surface for higher binding capacity for proteins and other                          molecules with both hydrophilic and hydrophobic regions.                          (Recommended for most ELISA experiments)▫ Medibinding : Surface adsorbs proteins and molecules that consists of interrmediate                          hydrophobic and hydrophilic properties.                          (Antigen/Antibody ELISA; More hydrophbic compared to Maxibinding Plates)▫ Unibinding    : Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) ▪ Strip single well type (8well/strip, breakable well)▪ Lid not included▪ Developed for immunoassay(ELISA) and general binding assay▪ Uniform surface homogeneity▪ Flat bottom▪ Raised rims on the wells▪ Alphanumeric labelling▪ Designed for easy handling and automation
Catalog No. 50-302-8985