Learn More
Medchemexpress LLC Immunoproteasome inhibitor 1 | 2755772-63-3 | 99.6% | 358.43 | C20H26N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1444525MG
Encompass_Preferred
Immunoproteasome inhibitor 1 is a small-molecule, reversible immunoproteasome inhibitor supplied as a high-purity solid for biochemical and cellular research. It is intended for experimental use in studies of proteasome function and immune-related proteostasis, with recommended storage to preserve activity.
  • High purity (>99.5%) suitable for analytical and biological assays.
  • Supplied as a solid for flexible formulation and handling.
  • Recommended storage conditions provided for stability and reproducibility.
  • Defined molecular formula and molecular weight for accurate dosing.
  • Provided with SMILES notation to support structure confirmation.
Catalog No. 50-002-21219