Learn More
IMPLEN INC Implen N60-Mobile Nano Photometer, Mirovolume (drop) UV/Vis Spectrophtometer with built-in battery pack and vortex
Supplier:  IMPLEN INC N60MOBILE
Encompass_Preferred
Implen is a leader in innovative, high-quality UV/VIS spectroscopy, and the NanoPhotometer trusted by thousands of labs worldwide. The NanoPhotometer N60 delivers full scan data from 200–900 nm in 2.5s for applications including quantification of DNA, RNA, proteins, kinetics, nanoparticles, and antibodies using only 0.3–2 µl drops. A recalibration-free design removes the need for costly service contracts, guaranteeing accuracy and stability without pedestal reconditioning. Advanced measurement technology eliminates innaccuracies from low surface tension and evaporation, ensuring reproducible results even with proteins or organic solvents. Sample Control™ and Blank Control™ enhance contaminant detection via ratios and spectral analysis, improving success of downstream applications such as (q)PCR, cloning, microarrays, drug production, and protein purification. Optional CFR21 software, IQ/OQ package, and REST API for LIMS integration make the N60 ideal for pharmaceutical labs
Catalog No. NC3385216
Only null left
Add to Cart
Request a Quote