Learn More
Selleck Chemical LLC Importazole 10mg 662163-81-7
SDP
Supplier:  Selleck Chemical LLC S844610MG
Encompass_Preferred
Importazole is a small molecule inhibitor of the transport receptor importin-? (Karyopherin beta) specifically blocking importin-?-mediated nuclear import. Importazole induces apoptosis and inhibits tumor growth. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6066