Learn More
Cayman Chemical Importazole 25mg
SDP
Supplier:  Cayman Chemical 2149125 MG
Encompass_Preferred
An inhibitor of importin-β-mediated nuclear import (IC50 = 15 μM in HEK293 cells); does not interfere with transportin-mediated nuclear import or chromosomal maintenance 1 (CRM1)-mediated nuclear export
Catalog No. 50-298-7051