Learn More
Medchemexpress LLC Impurity of Doxercalciferol | 127516-23-8 | C28H44O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY769375MG
Encompass_Preferred
Impurity of Doxercalciferol is an impurity of doxercalciferol, a synthetic analog of ergocalciferol (vitamin D2). It is used as an agent for secondary hyperparathyroidism and metabolic bone disease, and it suppresses parathyroid synthesis and secretion.
  • Used as an agent for secondary hyperparathyroidism
  • Used as an agent for metabolic bone disease
  • Suppresses parathyroid synthesis and secretion
  • Synthetic analog of ergocalciferol (vitamin D2)
  • Store at 4°C, protect from light, and under nitrogen
Catalog No. 50-004-33728