Learn More
Medchemexpress LLC HY-100431 5mg Medchemexpress, IMR-1 CAS:310456-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004315MG
Encompass_Preferred
Medchemexpress, HY-100431 5mg IMR-1 CAS:310456-65-6 IMR-1 is a novel class of Notch inhibitors targeting the transcriptional activation with an IC 50 of 26 μM. IMR-1 prevents the recruitment of Mastermind-like 1 (Maml1) to the Notch Ternary Complex (NTC) on chromatin, inhibits Notch target gene transcription and dramatically inhibits tumor growth in a patient-derived tumor xenograft model [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1831