Learn More
Optimize Technologies EXP™ Analytical Column In-Line Holder Kit, 50mm
SDP
Supplier:  Optimize Technologies 157004448
Encompass_Preferred
The cartridge-based EXP™ Analytical Column System is designed for superior chromatographic analysis with each injection rated for routine use up to 20,000 psi. The EXP™ Analytical Column Holders incorporate Free-Turn™ architecture allowing the user to quickly exchange columns by hand without breaking any fluid connections. The holders are available in two formats: In-Line and Direct Connect, allowing the user to bypass additional tubing and connect directly to the detector for even greater resolution. Holder kits include EXP™ Nuts and Titanium Hybrid Ferrules to ensure repeated zero-dead-volume connections. EXP™ Analytical Column Hardware is available for purchase as empty columns. The column hardware is assembled up to the point of packing, allowing the customer to completely control the resins used, packing processes and testing conditions. Packing fixtures are also available and are compatible with all column dimensions.
Catalog No. 50-200-6584