Learn More
Ocean Optics Inc INLINE-SFH Inline Filter Holder
Supplier:  Ocean Optics Inc INLINESFH
Encompass
The inline filter holder is a convenient sampling accessory that connects to optical fibers on either side of the holder. Users can measure optical filters from 10-12.7 mm diameter and up to 9 mm thickness by connecting one end of the holder to an illumination fiber (transmits light from a source) and the other to a read fiber (transmits light to the spectrometer).
Catalog No. NC1846067