Learn More
Sigma Aldrich Fine Chemicals Biosciences In Situ Cell Death Detecti
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11684817910
Encompass
Kit for the detection and quantification of apoptotic cell death on a single-cell level by light microscopy in immunohistocytochemistry.
Catalog No. 50-290-5201