Learn More
Medchemexpress LLC Inarigivir soproxil | 942123-43-5 | MFCD31715418 | 99.9% | 703.62 g/mol | C25H34N7O13PS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1090355MG
Encompass_Preferred
Inarigivir soproxil is an orally bioavailable antiviral prodrug used as a research reagent to evaluate innate immune agonism and antiviral activity against RNA viruses such as hepatitis B and C. It is supplied as a research-grade solid intended for laboratory experiments and not for human use.
  • Orally bioavailable prodrug for studying innate immune activation and antiviral effects.
  • High purity (99.87%) suitable for reliable experimental results.
  • Supplied as a white to off-white solid for straightforward handling.
  • Available in multiple small-vial sizes to support dose-ranging studies.
  • Store protected from light under nitrogen at -20°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-64558