Learn More
Selleck Chemical LLC INCB3344-5mg
SDP
Supplier:  Selleck Chemical LLC S82205MG
Encompass_Preferred
INCB3344 is a potent, selective and orally bioavailable CCR2 antagonist with IC50 values of 5.1 nM (hCCR2) and 9.5 nM (mCCR2) in binding antagonism and 3.8 nM (hCCR2) and 7.8 nM (mCCR2) in antagonism of chemotaxis activity.
Catalog No. 50-239-0026