Learn More
Darwin Chambers Company Single Door Thermoelectric Incubator 21 cu. ft.
SDP
Supplier:  Darwin Chambers Company TE030SS
Encompass_Preferred
The TE030-SS is a 23.9 cubic feet single door laboratory incubator. Temperature range is 18°C to 50°C. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The TE series chambers offer thermoelectric cooling that provides a much more stable temperature control than other refrigeration systems. All the TE Series incubators incorporate inherent cooling redundancy, quiet operation, simple serviceability and excellent reliability.
Catalog No. 50-189-3890
Only null left
Add to Cart
Request a Quote