Learn More
Sartorius INCUCYTE MS IGG1 FABFLUOR 594
Supplier:  Sartorius 4844
Encompass
Incucyte® Fabfluor-594 Antibody Labeling Dye is designed for quick, easy labeling of Fc- containing test antibodies with a green fluorophore. Once labeled the Incucyte Fabfluor-594-antibody complex, in combination with integrated Incucyte® Surface Fit background subtraction, can be used for identification of surface expressed antigens in live-cells. ESSEN BIOSCIENCE INC mfg# 4844
Catalog No. NC1935169
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov