Learn More
Sartorius Incucyte Nuclight Rapid Red Dye for Live-Cell Nuclear Labeling
Supplier:  Sartorius 4717
Encompass
Supplied as a single vial (50 µL), sufficient for 100 to 200 tests in a 96-well plate.
Catalog No. NC1404054
Add to Cart
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov