Learn More
MSC Partners Brand Industrial Aluminum Foil Tape, 5 mil, 1 Case
Supplier: MSC 39581871
This industrial aluminum foil tape is designed for demanding applications. It features a durable aluminum material and a strong acrylic adhesive, making it suitable for a variety of sealing, patching, and masking tasks in industrial environments.
- Material: aluminum
- Thickness: 5 mil
- Length: 60 yd.
- Width: 3 in
- Adhesive: acrylic
- Color: silver
- Grade: industrial
- Country of origin: United States
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.