Learn More
Medchemexpress LLC Indacrinone | 56049-88-8 | 365.21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYU002425MG
Encompass_Preferred
Indacrinone (MK196) is an investigational diuretic that exhibits pronounced saluretic activity in rats and dogs, and both uricosuric and saluretic activity in chimpanzees. It is intended for research use only.
  • Investigational diuretic for research
  • Displays strong saluretic and uricosuric activity
  • Powder stable for 3 years at -20°C
  • Solution stable for 6 months at -80°C or 1 month at -20°C
  • Highly soluble in DMSO
Catalog No. 50-004-49690