Learn More
Selleck Chemical LLC Indapamide 50mg 26807-65-8
SDP
Supplier:  Selleck Chemical LLC S173050MG
Encompass_Preferred
Indapamide is a non-thiazide sulphonamide diuretic compound, generally used in the treatment of hypertension, as well as decompensated cardiac failure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-681-1