Learn More
TARGETMOL CHEMICALS INC Indecainide 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T321551MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Indecainide (Ricainid) is a novel antiarrhythmic compound that is carcinogenic and can be used to study chronic stable ventricular arrhythmias secondary to coronary artery disease or cardiomyopathy. Purity 98.13%
Catalog No. 50-287-6325