Learn More
Hampton Research HR2-944-19/Index #19 (B7), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 8.2
SDP
Supplier:  Hampton Research HR294419
Encompass
HR2-944-19/Index #19 (B7), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 8.2. Please note items from this supplier are non-returnable.
Catalog No. 50-219-2