Learn More
Hampton Research HR2-944-30/Index #30 (C6), 185 ml - 0.1 M Sodium chloride, 0.1 M BIS-TRIS pH 6.5, 1.5 M Ammonium sulfate
SDP
Supplier:  Hampton Research HR294430
Encompass
HR2-944-30/Index #30 (C6), 185 ml - 0.1 M Sodium chloride, 0.1 M BIS-TRIS pH 6.5, 1.5 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-220-3