Learn More
Hampton Research HR2-944-51/Index #51 (E03), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 6.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol
SDP
Supplier:  Hampton Research HR294451
Encompass
HR2-944-51/Index #51 (E03), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 6.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-222-4