Learn More
Grainger Indexable Toolholder SCBC HSS
Supplier:  Grainger 16R836
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R836 is a right-hand turning toolholder with a 1" shank diameter and a 0.75" insert size. It has a positive rake angle of 5 degrees and accepts CCGT and CCMT style inserts. The toolholder body is made from steel and has an overall length of 5". It can be used for turning, facing, profiling and grooving applications on lathes and CNC lathes. The toolholder allows for precision machining with indexable carbide inserts.
Catalog No. 50-276-6639