Learn More
Grainger Indexable Toolholder SCBC HSS
Supplier:  Grainger 16R837
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R837 is a right-hand turning toolholder for indexable inserts. It has a shank diameter of 1 inch, an overall length of 5 inches, and accepts triangular inserts with a thickness of 0.375 inches. The toolholder is made from steel and has a black oxide finish. It can be used for turning, facing, profiling and grooving operations. The toolholder allows for precision machining with indexable inserts, providing efficient material removal and a quality surface finish. It is suitable for use on CNC lathes and manual engine lathes. The toolholder requires an ANSI standard triangular insert for complete operation.
Catalog No. 50-276-6934