Learn More
Grainger Indexable Toolholder SDNC HSS
Supplier:  Grainger 16T409
Encompass_Preferred
The Micro 100 Indexable Turning Toolholder 16R846 is a right-hand turning toolholder with a 16mm shank diameter and a 0.787 inch insert size. It has a positive rake angle of 7 degrees and accepts CCGT and CCMT style inserts. The toolholder body is made from steel and has a black oxide finish. It is designed for light to medium duty turning applications. The toolholder allows for precision turning while minimizing vibration. It accepts triangular, square, and round inserts, providing versatility. The toolholder can be used on most lathes and is a quality option for a wide range of materials.
Catalog No. 50-276-8343