Learn More
Medchemexpress LLC Indibulin | 204205-90-3 | MFCD05861105 | 99.9% | 389.83 g/mol | C22H16ClN3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY136495MG
Encompass_Preferred
Indibulin is an orally active inhibitor of tubulin assembly used in research to study microtubule dynamics and anticancer effects. It disrupts spindle formation, activates mitotic checkpoint proteins, and induces mitotic arrest and apoptosis.
  • Orally active tubulin assembly inhibitor used for anticancer research.
  • Demonstrates potent anticancer activity with minimal neurotoxicity.
  • High purity solid suitable for biochemical and cell-based assays.
  • Soluble in DMSO (50 mg/mL) for in vitro use; may require sonication.
  • Formulatable for in vivo studies in DMSO/corn oil and DMSO/SBE-β-CD with reported solubilities.
  • Stable as powder when stored at -20°C for long-term storage.
Catalog No. 50-002-21841