Learn More
Cayman Chemical INDIGlo LucIfRase detect 50mL
SDP
Supplier:  Cayman Chemical 3649550 ML
Encompass_Preferred
INDIGlo Luciferase Detection Reagent (LDR) is formulated to deliver steady, long-lived luminescence signal from mammalian reporter cells expressing firefly luciferase.
Catalog No. 50-291-2902