Learn More
Indigo Biosciences INDIGO Biosciences Assay, Tropomysin Receptor Kinase C (TrkC; NTRK3), Human, 384-Well Format
Supplier:  Indigo Biosciences IB27032
Encompass_Preferred
This cell-based reporter assay is designed for the detection of Tropomysin Receptor Kinase C activity. It provides a complete system for screening and evaluating compounds that modulate TrkC receptor signaling.
  • Designed for human TrkC.
  • Provided in a convenient 384-well assay format.
  • Offers reliable and reproducible results for in vitro toxicology solutions.
  • Accelerates scientific decision-making in nuclear receptor and in vitro toxicology research.
Catalog No. NC3978862