Learn More
INDICAL INC IndiMag 48 PW Rod cover (672 reactions)
SDP
Supplier:  INDICAL INC PW940237
Encompass_Preferred
The IndiMag 48 PW Rod Cover (672 reactions) includes 84 strips, with each strip designed to process 8 samples. This set is compatible with the IndiMag 2 or IndiMag 48 automated DNA/RNA magnetic rod extraction devices and supports a total of 672 reactions/samples. Each 8-well rod cover securely locks into position within the instrument’s processing chamber. Optimized for magnetic bead processing, the tip combs ensure optimal recovery of magnetic beads, delivering excellent performance throughout the extraction process.
Catalog No. NC3451755