Learn More
Cayman Chemical IndIrubIn-3-monxIme 1mg
SDP
Supplier:  Cayman Chemical 133141 MG
Encompass_Preferred
A potent inhibitor of GSK3β (IC50 = 22 nM), preventing tau phosphorylation both in vitro and in vivo
Catalog No. 50-295-2545