Learn More
Medkoo Biosciences Inc Indisulam, 165668-41-7, MFCD00945325, 25mg
SDP
Supplier:  Medkoo Biosciences Inc 20154025MG
Encompass
Chemical Formula: C14H12ClN3O4S2, Purity:>98%, Molecular Weight: 385.84, Soluble in DMSO, not in water. Indisulam, also known as E7070, is a carbonic anhydrase inhbitor, is also a novel synthetic sulfonamide that targets the G1 phase of the cell cycle.
Catalog No. NC1369954