Learn More
Strem, An Ascensus Company CAS# 13465-10-6. 2g. Indium(I) chloride, anhydrous (99.99%-In) PURATREM. MFCD00148875
SDP
Supplier:  Strem, An Ascensus Company 9349302G
Encompass
CAS# 13465-10-6. 2g. Indium(I) chloride, anhydrous (99.99%-In) PURATREM. MFCD00148875. Molecular Weight: 150.27. Molecular Formula: InCl. Color/form: golden yellow pwdr. Strem# 93-4930. http://www.strem.com/catalog/v/93-4930/
Catalog No. 50-901-15520