Learn More
Strem, An Ascensus Company CAS# 22519-64-8. 25g. Indium(III) chloride tetrahydrate (99.99%-In) PURATREM. MFCD00149746
SDP
Supplier:  Strem, An Ascensus Company 93493125G
Encompass
CAS# 22519-64-8. 25g. Indium(III) chloride tetrahydrate (99.99%-In) PURATREM. MFCD00149746. Molecular Weight: 221.18 (293.26). Molecular Formula: InCl3∙4H2O. Color/form: white xtl. Strem# 93-4931. http://www.strem.com/catalog/v/93-4931/
Catalog No. 50-901-15523