Learn More
Strem, An Ascensus Company CAS# 13510-35-5. 1g. Indium(III) iodide (99.999%-In) PURATREM. MFCD00016151
SDP
Supplier:  Strem, An Ascensus Company 9349151G
Encompass
CAS# 13510-35-5. 1g. Indium(III) iodide (99.999%-In) PURATREM. MFCD00016151. Molecular Weight: 495.53. Molecular Formula: InI3. Color/form: yellow to red solid Strem# 93-4915. http://www.strem.com/catalog/v/93-4915/
Catalog No. 50-901-15507