Learn More
Medchemexpress LLC Indium(III) isopropoxide | 38218-24-5 | MFCD00210318 | 292.08 g/mol | C9H21InO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1330235MG
Encompass_Preferred
Indium(III) isopropoxide is an organometallic indium reagent supplied as a solid. It is used as a Lewis acid and hydrogen-transfer catalyst in organic synthesis and serves as a precursor for indium oxide thin films and nanomaterials. The compound is provided in small research quantities for laboratory use.
  • Acts as a Lewis acid catalyst in organic transformations.
  • Functions as a hydrogen-transfer catalyst for selective reductions.
  • Serves as a precursor for deposition of indium oxide and related materials.
  • Supplied as a solid in small, research-scale quantities suitable for lab work.
  • Compatible with typical organic synthesis workflows and characterization methods.
Catalog No. 50-002-62520
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.