Learn More
Strem, An Ascensus Company CAS# 1312-43-2. 5g. Indium(III) oxide (99.998%-In) PURATREM. MFCD00011060
SDP
Supplier:  Strem, An Ascensus Company 9349065G
Encompass
CAS# 1312-43-2. 5g. Indium(III) oxide (99.998%-In) PURATREM. MFCD00011060. Molecular Weight: 277.64. Molecular Formula: In2O3. Color/form: yellow pwdr. Strem# 93-4906. http://www.strem.com/catalog/v/93-4906/
Catalog No. 50-901-15503