Learn More
Chemglass Life Sciences Chemglass Chem-Spin Rotary Scraper Tool, Individual - Fits 100 mL Round Bottom Flasks (CG-1981-C100)
SDP
Supplier:  Chemglass Life Sciences CG1981C100
Encompass_Preferred
The Chem-Spin rotary scraper is a motor-driven scraping tool designed to remove dried solids from the interior walls of 100 mL round bottom flasks quickly and with minimal effort. Constructed from stainless steel and PTFE, the tool is laser-etched for clear identification and designed to work with standard laboratory joints.
  • Removes dried solids from flask walls with minimal manual effort.
  • Constructed of stainless steel and PTFE for chemical resistance.
  • Designed to fit 100 mL round bottom flasks.
  • Compatible with 24/40 or larger standard taper joints.
  • Laser-etched size and identification for easy inventory control.
Catalog No. 50-273-3540