Learn More
Chemglass Life Sciences Chem-Spin individual rotary scraper tool, fits 500 mL round-bottom flask
SDP
Supplier:  Chemglass Life Sciences CG1981C500
Encompass_Preferred
Easily remove dried-on solids from the interior walls of 500 mL round-bottom flasks without scratching or harming the glass. The motor-driven rotary scraper brushes the flask surface and collects loose solids at the bottom, eliminating the need for manual scraping.
  • Easily removes dried-on solids without scratching the flask.
  • Designed for sweeping action across the full radius of the flask bulb.
  • Fits 500 mL round-bottom flasks and fits outer joint size 24/40 or larger.
  • Constructed of stainless steel with PTFE components for chemical resistance.
  • Sold as an individual replacement tool for rotary scraper systems.
Catalog No. 50-273-3539