Learn More
Medchemexpress LLC HY-18763 5mg Medchemexpress, Indobufen CAS:63610-08-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187635MG
Encompass_Preferred
Medchemexpress, HY-18763 5mg Indobufen CAS:63610-08-2 Indobufen is a platelet aggregation inhibitor. Indobufen is a reversible platelet cyclooxygenase (Cox) activity inhibitor. Indobufen suppresses thromboxane A2 (TxA2) synthesis. Indobufen down-regulates tissue factor (TF) in monocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0014