Learn More
Medchemexpress LLC Indobufen | 63610-08-2 | 99.98% | 295.33 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1876350MG
Encompass_Preferred
Indobufen is a platelet aggregation inhibitor and a reversible platelet cyclooxygenase (Cox) activity inhibitor. It suppresses thromboxane A2 (TxA2) synthesis and down-regulates tissue factor (TF) in monocytes. This compound is intended for research and analytical applications, serving as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Platelet aggregation inhibitor
  • Reversible platelet cyclooxygenase (Cox) activity inhibitor
  • Suppresses thromboxane A2 (TxA2) synthesis
  • Down-regulates tissue factor (TF) in monocytes
  • Analytical standard for research applications
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-88215