Learn More
Medchemexpress LLC Indole | 120-72-9 | 117.15 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW001132500MG
Encompass_Preferred
Indole is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. It regulates bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence, acting as an intercellular signal molecule.
  • Aromatic, heterocyclic, organic compound
  • Widely distributed in the natural environment
  • Produced by various bacteria
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule in bacteria
  • Supports bacterial spore formation, plasmid stability, drug resistance, biofilm formation, and virulence
Catalog No. 50-004-46625