Learn More
Medchemexpress LLC Indole-13C | 85597-55-3 | 118.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW001132S150MG
Encompass_Preferred
Indole-13C is the 13C labeled Indole, an endogenous metabolite. It is intended for research use only and not for patient use. This compound serves as a tracer and internal standard for quantitative analysis in various analytical techniques.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotope of carbon
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-49173