Learn More
TARGETMOL CHEMICALS INC Indole-3-acetamide 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T48341G
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Indole-3-acetamide is a biosynthetic intermediate of indole-3-acetic acid. Indole-3-acetamide is a biosynthetic intermediate of indole-3-acetic acid, which is the most common natural plant growth hormone in the phytogrowth factor class. Purity 99.86%
Catalog No. 50-287-8681