Learn More
Medchemexpress LLC Indole-3-acetamide | 879-37-8 | MFCD00005641 | >98.0% | 174.20 | C10H10N2O | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0167845G
Encompass_Preferred
Indole-3-acetamide is an endogenous indole metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). It is supplied as an analytical standard for biochemical and plant-biology research, with documented solubility profiles and recommended storage conditions for powdered and solvent solutions.
  • Contains CAS 879-37-8; formula C10H10N2O; molecular weight 174.20.
  • High solubility in DMSO (≈100 mg/mL) and moderate solubility in water (≈1.3 mg/mL).
  • Forms clear solutions in DMSO, water, PBS, and corn oil.
  • Powder stable at -20°C for long-term storage; in solution store at -80°C to maximize stability.
  • Suitable as an analytical standard for assays and studies of auxin biosynthesis.
  • Solvent preparation examples provided for 1 mg, 5 mg, and 10 mg quantities.
Catalog No. 50-002-99865