Learn More
Cayman Chemical Indole-3-acetIc AcId-d5 25mg
SDP
Supplier:  Cayman Chemical 3889425 MG
Encompass_Preferred
An internal standard for the quantification of indole-3-acetic acid by GC- or LC-MS
Catalog No. 50-291-8929