Learn More
Medchemexpress LLC Indole-3-acetic acid-d7 | 1173020-21-7 | MFCD04118278 | 99.8% | 182.23 g/mol | C10H2D7NO2 | 10 MG

Supplier: Medchemexpress LLC HY18569S310MG
3-Indoleacetic acid-d7 is a deuterium-labeled isotopic analogue of indole-3-acetic acid used as an internal standard and research reagent for analytical and plant biology applications. It provides a consistent mass shift for accurate quantitation in mass spectrometry and chromatographic assays.
- Deuterium-labeled internal standard (d7) for analytical quantitation.
- High purity: 99.84%.
- Chemical formula: C10H2D7NO2.
- Molecular weight: 182.23 g/mol.
- Supplied in 10 mg vials for calibration and method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.