Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetyl β-D-glucopyranose | 19817-95-9 | 337.32 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN130141MG

Encompass_Preferred

Indole-3-acetyl β-D-glucopyranose is a glucose esterified derivative of the plant hormone indole-3-acetic acid (IAA). It is involved in regulating plant growth and can be used to study the metabolism and regulatory mechanisms of plant hormones. This product is for research use only.

  • Glucose esterified derivative of indole-3-acetic acid (IAA)
  • Regulates plant growth
  • Used to study plant hormone metabolism and regulatory mechanisms
  • High purity: 98.72%
  • Soluble in water: 150 mg/mL (requires ultrasonic)
  • Available documentation includes data sheet, SDS, COA, HNMR, RP-HPLC, and MS
  • Recommended storage at -20°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-16189


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.