Learn More
Medchemexpress LLC Indole-3-acetyl β-D-glucopyranose | 19817-95-9 | 337.32 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN130145MG
Encompass_Preferred
Indole-3-acetyl β-D-glucopyranose is a glucose esterified derivative of the plant hormone indole-3-acetic acid (IAA). It is involved in regulating plant growth and can be used to study the metabolism and regulatory mechanisms of plant hormones.
  • Involved in regulating plant growth.
  • Can be used to study the metabolism and regulatory mechanisms of plant hormones.
  • Purity: 98.72%.
Catalog No. 50-004-16190